Assay Details
| Gene Symbol: Lrrc28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027413.1 | NP_081689.1 | 5 - 6 | 509 | 112 | |
| GenBank mRNA | AK008713.1 | - | 5 - 6 | 477 | 112 | |
| AK050154.1 | - | 5 - 6 | 481 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc28
Genomic Map