Assay Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC027062.1 | - | 1 - 2 | 335 | 5355240 | 76 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027062.1 transcripts.
Set Membership: |
|

Peli2
Genomic Map