Assay Details
| Gene Symbol: 2210411M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008903.1 | - | 1 - 2 | 166 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 63a | Tmem63a |
BC014795, Tmem64a |
Anti-sense | Chr. 1 - 180941225 - 180975106 on NCBI Build GRCm38 | |
| Downstream |
epoxide hydrolase 1, microsomal |
Ephx1 |
AI195553, Eph-1, Eph1, Epxh1, mEH |
31.4kb | Sense | Chr. 1 - 180989556 - 181017495 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

2210411M09Rik
Genomic Map