Assay Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290278.1 | NP_001277207.1 | 2 - 3 | 361 | 66 | |
| NM_001290279.1 | NP_001277208.1 | 2 - 3 | 361 | 66 | ||
| NM_001290280.1 | NP_001277209.1 | 2 - 3 | 361 | 66 | ||
| NM_001290282.1 | NP_001277211.1 | 2 - 3 | 361 | 66 | ||
| NM_144814.3 | NP_659063.3 | 2 - 3 | 361 | 66 | ||
| GenBank mRNA | AK040839.1 | - | 2 - 3 | 348 | 66 | |
| AK135612.1 | - | 2 - 3 | 334 | 66 | ||
| AK155440.1 | - | 2 - 3 | 333 | 66 | ||
| BC020065.1 | - | 2 - 3 | 248 | 66 | ||
| BC057141.1 | - | 2 - 3 | 108 | 5151580 | 66 | |
| BC138938.1 | - | 2 - 3 | 81 | 8861960 | 66 | |
| BC138939.1 | - | 2 - 3 | 81 | 8861961 | 66 | |
| BC145469.1 | - | 2 - 3 | 81 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290282.1 transcripts.
Set Membership: |
|

Rcor3
Genomic Map