Assay Details
| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029987.2 | NP_084263.2 | 5 - 6 | 549 | 68 | |
| GenBank mRNA | AF410461.1 | - | 4 - 5 | 497 | 68 | |
| AK020906.1 | - | 5 - 6 | 548 | 68 | ||
| BC090656.1 | - | 5 - 6 | 525 | 68 | ||
| BC130028.1 | - | 4 - 5 | 501 | 40140883 | 68 | |
| CJ133618.1 | - | 5 - 6 | 550 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpe65
Genomic Map