Assay Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB095735.1 | - | 13 - 14 | 1369 | 83 | |
| AB095736.1 | - | 13 - 14 | 1369 | 83 | ||
| AK054412.1 | - | 13 - 14 | 1359 | 83 | ||
| AK133177.1 | - | 13 - 14 | 1368 | 83 | ||
| AK161401.1 | - | 10 - 11 | 970 | 83 | ||
| AK164249.1 | - | 13 - 14 | 1369 | 83 | ||
| AK166351.1 | - | 13 - 14 | 1377 | 83 | ||
| BC060208.1 | - | 13 - 14 | 1328 | 83 | ||
More Information
Set Membership: |
|

Dtl
Genomic Map