Assay Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB095735.1 | - | 4 - 5 | 452 | 104 | |
| AB095736.1 | - | 4 - 5 | 452 | 104 | ||
| AK012919.1 | - | 4 - 5 | 453 | 104 | ||
| AK054412.1 | - | 4 - 5 | 442 | 104 | ||
| AK133177.1 | - | 4 - 5 | 451 | 104 | ||
| AK154594.1 | - | 4 - 5 | 460 | 104 | ||
| AK164249.1 | - | 4 - 5 | 451 | 104 | ||
| AK166351.1 | - | 4 - 5 | 460 | 104 | ||
| AK169601.1 | - | 4 - 5 | 451 | 104 | ||
| BC060208.1 | - | 4 - 5 | 412 | 104 | ||
| CF898098.1 | - | 4 - 5 | 474 | 104 | ||
More Information
Set Membership: |
|

Dtl
Genomic Map