Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 10 - 11 | 1442 | 87 | |
| GenBank mRNA | AK028896.1 | - | 10 - 11 | 1405 | 87 | |
| AK030331.1 | - | 9 - 10 | 1442 | 87 | ||
| AK134973.1 | - | 9 - 10 | 1373 | 87 | ||
| AK145570.1 | - | 10 - 11 | 1443 | 87 | ||
| AK154969.1 | - | 9 - 10 | 1223 | 87 | ||
| AK173249.1 | - | 10 - 11 | 1390 | 87 | ||
| DQ073331.1 | - | 9 - 10 | 1198 | 87 | ||
| X85992.1 | - | 8 - 9 | 1034 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4b
Genomic Map