Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 1 - 2 | 148 | 77 | |
| GenBank mRNA | AK028896.1 | - | 1 - 2 | 111 | 77 | |
| AK145570.1 | - | 1 - 2 | 149 | 77 | ||
| AK173249.1 | - | 1 - 2 | 96 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145570.1, NM_013659.4 transcripts.
Set Membership: |
|

Sema4b
Genomic Map