Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 5 - 6 | 734 | 112 | |
| GenBank mRNA | AK028896.1 | - | 5 - 6 | 697 | 112 | |
| AK030331.1 | - | 4 - 5 | 734 | 112 | ||
| AK145570.1 | - | 5 - 6 | 735 | 112 | ||
| AK154969.1 | - | 4 - 5 | 515 | 112 | ||
| AK173249.1 | - | 5 - 6 | 682 | 112 | ||
| DQ073331.1 | - | 4 - 5 | 490 | 112 | ||
| X85992.1 | - | 3 - 4 | 326 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4b
Genomic Map