Assay Details
| Gene Symbol: Ifi44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ299405.1 | - | 2 - 3 | 552 | 78 | |
| BC111882.1 | - | 2 - 3 | 552 | 40060245 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifi44
Genomic Map