Assay Details
| Gene Symbol: Ifi44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133871.2 | NP_598632.2 | 6 - 7 | 1179 | 82 | |
| GenBank mRNA | AJ299405.1 | - | 5 - 6 | 1069 | 82 | |
| AK079749.1 | - | 6 - 7 | 1131 | 82 | ||
| AK136553.1 | - | 6 - 7 | 1131 | 82 | ||
| AK151346.1 | - | 6 - 7 | 1130 | 82 | ||
| AK171676.1 | - | 6 - 7 | 1112 | 82 | ||
| BC026901.1 | - | 1 - 2 | 77 | 82 | ||
| BC111882.1 | - | 5 - 6 | 1069 | 40060245 | 82 | |
| BC112402.1 | - | 5 - 6 | 1069 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifi44
Genomic Map