Assay Details
| Gene Symbol: Abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133722.2 | NP_598483.2 | 1 - 2 | 608 | 106 | |
| GenBank mRNA | BC018511.1 | - | 1 - 2 | 608 | 3483379 | 106 |
| BM943880.1 | - | 1 - 2 | 89 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abhd17c
Genomic Map