Assay Details
| Gene Symbol: Dhrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027790.2 | NP_082066.2 | 3 - 4 | 220 | 85 | |
| GenBank mRNA | AK017274.1 | - | 3 - 4 | 222 | 85 | |
| BC117731.2 | - | 2 - 3 | 113 | 40089799 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017274.1, BC117731.2, NM_027790.2 transcripts.
Set Membership: |
|

Dhrs2
Genomic Map