Assay Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146055.3 | NP_666167.3 | 7 - 8 | 780 | 90 | |
| GenBank mRNA | AK028194.1 | - | 7 - 8 | 702 | 90 | |
| AK031472.1 | - | 3 - 4 | 320 | 90 | ||
| AK050238.1 | - | 7 - 8 | 780 | 90 | ||
| BC021475.1 | - | 7 - 8 | 810 | 5343456 | 90 | |
| BC038346.1 | - | 7 - 8 | 732 | 3985250 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031472.1 transcripts.
Set Membership: |
|

Ccar2
Genomic Map