Assay Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133809.1 | NP_598570.1 | 4 - 5 | 330 | 115 | |
| GenBank mRNA | AK129011.1 | - | 343 | 115 | ||
| AK133074.1 | - | 438 | 115 | |||
| AK158251.1 | - | 4 - 5 | 526 | 115 | ||
| AK171812.1 | - | 4 - 5 | 454 | 115 | ||
| BC014683.1 | - | 4 - 5 | 330 | 4210278 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kmo
Genomic Map