Assay Details
| Gene Symbol: Mttp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163457.1 | NP_001156929.1 | 15 - 16 | 2254 | 85 | |
| NM_008642.2 | NP_032668.2 | 13 - 14 | 1944 | 85 | ||
| GenBank mRNA | AK146553.1 | - | 13 - 14 | 1945 | 85 | |
| BC012686.1 | - | 13 - 14 | 1904 | 4163901 | 85 | |
| EU553486.1 | - | 13 - 14 | 1912 | 85 | ||
| L47970.1 | - | 13 - 14 | 1891 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mttp
Genomic Map