Assay Details
| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277944.1 | NP_001264873.1 | 3 - 4 | 306 | 64 | |
| GenBank mRNA | AK146468.1 | - | 2 - 3 | 107 | 64 | |
| BC024697.1 | - | 3 - 4 | 306 | 4195301 | 64 | |
| BG085960.2 | - | 2 - 3 | 149 | 64 | ||
| CB949564.1 | - | 2 - 3 | 212 | 64 | ||
| Z15090.1 | - | 2 - 3 | 107 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apoc2
Genomic Map