Assay Details
| Gene Symbol: Pla2g12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286948.1 | NP_001273877.1 | 1 - 2 | 328 | 83 | |
| NM_023196.4 | NP_075685.2 | 1 - 2 | 328 | 83 | ||
| NR_104611.1 | 1 - 2 | 328 | 83 | |||
| GenBank mRNA | AK003183.1 | - | 1 - 2 | 288 | 83 | |
| AK009133.1 | - | 1 - 2 | 330 | 83 | ||
| AK010011.1 | - | 1 - 2 | 318 | 83 | ||
| AK010174.1 | - | 1 - 2 | 294 | 83 | ||
| AK012414.1 | - | 1 - 2 | 256 | 83 | ||
| AK163693.1 | - | 1 - 2 | 242 | 83 | ||
| AK178670.1 | - | 1 - 2 | 36 | 83 | ||
| AY007381.1 | - | 1 - 2 | 301 | 83 | ||
| BC026812.1 | - | 1 - 2 | 293 | 4456431 | 83 | |
| BC051117.1 | - | 1 - 2 | 328 | 6306803 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g12a
Genomic Map