Assay Details
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145964.2 | NP_666076.2 | 6 - 7 | 646 | 85 | |
| GenBank mRNA | BB611022.1 | - | 6 - 7 | 334 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1ar
Genomic Map