Assay Details
| Gene Symbol: Rgma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177740.5 | NP_808408.2 | 1 - 2 | 365 | 107 | |
| GenBank mRNA | BC059072.1 | - | 1 - 2 | 365 | 6826181 | 107 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059072.1, NM_177740.5 transcripts.
Set Membership: |
|

Rgma
Genomic Map