Assay Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029585.3 | NP_083861.1 | 2 - 3 | 1195 | 68 | |
| GenBank mRNA | AK011666.1 | - | 2 - 3 | 1221 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Det1
Genomic Map