Assay Details
| Gene Symbol: A930011O12Rik | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040709.1 | 4 | 71 | ||||
| GenBank mRNA | AK044422.1 | - | LIT3369 RNCR3 |
4 | 71 | ||
| BC030402.1 | - | 2 | 71 | ||||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930011O12Rik
Genomic Map