Assay Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011183.3 | NP_035313.2 | 1 - 2 | 93 | 58 | |
| GenBank mRNA | BY136632.1 | - | 1 - 2 | 93 | 58 | |
| U57324.1 | - | 1 - 2 | 44 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011183.3, U57324.1 transcripts.
Set Membership: |
|

Psen2
Genomic Map