Assay Details
| Gene Symbol: Itpkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081175.1 | NP_001074644.1 | 5 - 6 | 2941 | 141 | |
| GenBank mRNA | AK039369.1 | - | 4 - 5 | 856 | 141 | |
| BC151160.1 | - | 5 - 6 | 2807 | 9088049 | 141 | |
| BC157986.1 | - | 5 - 6 | 2806 | 9088142 | 141 | |
| BC158000.1 | - | 5 - 6 | 2806 | 9088148 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itpkb
Genomic Map