Assay Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033285.1 | NP_001028457.1 | 31 - 32 | 4490 | 132 | |
| GenBank mRNA | AK129148.1 | - | 2 - 3 | 305 | 132 | |
| BC096679.1 | - | 1 - 2 | 82 | 132 | ||
| BC158017.1 | - | 31 - 32 | 4490 | 9007323 | 132 | |
| BC158095.1 | - | 30 - 31 | 4451 | 9007382 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc42bpa
Genomic Map