Assay Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022023.2 | NP_071306.2 | 5 - 6 | 425 | 100 | |
| GenBank mRNA | AB001732.1 | - | 5 - 6 | 357 | 100 | |
| AB050013.1 | - | 5 - 6 | 367 | 100 | ||
| AF297220.1 | - | 5 - 6 | 403 | 100 | ||
| AK013947.1 | - | 5 - 6 | 382 | 100 | ||
| AK013970.1 | - | 5 - 6 | 378 | 100 | ||
| AK142014.1 | - | 5 - 6 | 397 | 100 | ||
| BC040233.1 | - | 5 - 6 | 359 | 5364203 | 100 | |
| BC049134.1 | - | 5 - 6 | 366 | 6831501 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmfb
Genomic Map