Assay Details
| Gene Symbol: Tnfrsf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164155.1 | NP_001157627.1 | 1 - 2 | 140 | 115 | |
| GenBank mRNA | AB040432.1 | - | 1 - 2 | 104 | 115 | |
| CB244999.1 | - | 1 - 2 | 139 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB040432.1, NM_001164155.1 transcripts.
Set Membership: |
|

Tnfrsf19
Genomic Map