Assay Details
| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163836.1 | NP_001157308.1 | 1 | 1520 | 81 | |
| GenBank mRNA | AK016578.1 | - | 1 | 1513 | 81 | |
| AK029765.1 | - | 1 | 1501 | 81 | ||
| BC113177.1 | - | 1 | 1490 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pabpc6
Genomic Map