Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190336.1 | NP_001177265.1 | 2 - 3 | 142 | 78 | |
| NM_001190338.1 | NP_001177267.1 | 1 - 2 | 138 | 78 | ||
| GenBank mRNA | AB009369.1 | - | 1 - 2 | 141 | 78 | |
| BC100758.1 | - | 2 - 3 | 138 | 40046794 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB009369.1, BC100758.1, NM_001190336.1 transcripts.
Set Membership: |
|

Ccr6
Genomic Map