Assay Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177591.4 | NP_808259.2 | 10 - 11 | 1844 | 110 | |
| NM_183335.2 | NP_899178.2 | 9 - 10 | 1708 | 110 | ||
| NM_183336.2 | NP_899179.1 | 1 - 2 | 98 | 110 | ||
| GenBank mRNA | AK030701.1 | - | 1 - 2 | 100 | 110 | |
| AK129126.1 | - | 10 - 11 | 1826 | 110 | ||
| AY227771.1 | - | 10 - 11 | 1838 | 110 | ||
| AY227773.1 | - | 9 - 10 | 1708 | 110 | ||
| AY227774.1 | - | 1 - 2 | 93 | 110 | ||
| BC119264.1 | - | 1 - 2 | 61 | 8734013 | 110 | |
| BC119266.1 | - | 1 - 2 | 61 | 8734015 | 110 | |
| BC144762.1 | - | 1 - 2 | 61 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY227774.1, BC119264.1, BC119266.1, NM_183336.2 transcripts.
Set Membership: |
|

Igsf1
Genomic Map