Assay Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177591.4 | NP_808259.2 | 11 - 12 | 1875 | 76 | |
| NM_183335.2 | NP_899178.2 | 10 - 11 | 1739 | 76 | ||
| NM_183336.2 | NP_899179.1 | 2 - 3 | 129 | 76 | ||
| GenBank mRNA | AK030701.1 | - | 2 - 3 | 131 | 76 | |
| AK129126.1 | - | 11 - 12 | 1857 | 76 | ||
| AY227771.1 | - | 11 - 12 | 1869 | 76 | ||
| AY227773.1 | - | 10 - 11 | 1739 | 76 | ||
| AY227774.1 | - | 2 - 3 | 124 | 76 | ||
| BC119264.1 | - | 2 - 3 | 92 | 8734013 | 76 | |
| BC119266.1 | - | 2 - 3 | 92 | 8734015 | 76 | |
| BC144762.1 | - | 2 - 3 | 92 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf1
Genomic Map