Assay Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177591.4 | NP_808259.2 | 4 - 5 | 533 | 84 | |
| NM_177915.4 | NP_808583.1 | 4 - 5 | 533 | 84 | ||
| NM_183335.2 | NP_899178.2 | 3 - 4 | 397 | 84 | ||
| GenBank mRNA | AK030452.1 | - | 4 - 5 | 505 | 84 | |
| AK129126.1 | - | 4 - 5 | 515 | 84 | ||
| AY227771.1 | - | 4 - 5 | 527 | 84 | ||
| AY227772.1 | - | 4 - 5 | 527 | 84 | ||
| AY227773.1 | - | 3 - 4 | 397 | 84 | ||
| BC089474.1 | - | 4 - 5 | 514 | 84 | ||
| BC100420.1 | - | 4 - 5 | 533 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf1
Genomic Map