Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011076.2 | NP_035206.2 | 1 - 2 | 129 | 72 | |
| GenBank mRNA | AK008292.1 | - | 1 - 2 | 130 | 72 | |
| M30697.1 | - | 1 - 2 | 143 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008292.1, M30697.1, NM_011076.2 transcripts.
Set Membership: |
|

Abcb1a
Genomic Map