Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011076.2 | NP_035206.2 | 4 - 5 | 411 | 91 | |
| GenBank mRNA | AY864315.1 | - | 3 - 4 | 274 | 91 | |
| M30697.1 | - | 4 - 5 | 425 | 91 | ||
| M33581.1 | - | 4 - 5 | 410 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map