Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011076.2 | NP_035206.2 | 7 - 8 | 827 | 79 | |
| GenBank mRNA | AY864315.1 | - | 6 - 7 | 690 | 79 | |
| M24417.1 | - | 1 - 2 | 176 | 79 | ||
| M30697.1 | - | 7 - 8 | 841 | 79 | ||
| M33581.1 | - | 7 - 8 | 826 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map