Assay Details
| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025832.2 | NP_080108.1 | 10 - 11 | 1147 | 148 | |
| GenBank mRNA | AK005056.1 | - | 10 - 11 | 1157 | 148 | |
| AK007755.1 | - | 10 - 11 | 1183 | 148 | ||
| AK028432.1 | - | 10 - 11 | 1258 | 148 | ||
| AK152498.1 | - | 10 - 11 | 1157 | 148 | ||
| BC052445.1 | - | 10 - 11 | 1147 | 6412745 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa16
Genomic Map