| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
NM_001112703.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
NM_001283045.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
NM_001283046.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
NM_001283047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
NM_009594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004835.mm.1 |
TC0200000558.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
NM_001112703.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
NM_001283045.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
NM_001283046.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
NM_001283047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
NM_009594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004836.mm.1 |
TC0200000558.mm.1 |
MTA TC |