Assay Details
| Gene Symbol: Swi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175190.5 | NP_780399.1 | 1 - 2 | 184 | 58 | |
| NR_110965.1 | 1 - 2 | 184 | 58 | |||
| GenBank mRNA | AK013517.1 | - | 1 - 2 | 144 | 58 | |
| AK018775.1 | - | 1 - 2 | 122 | 58 | ||
| AK031907.1 | - | 1 - 2 | 152 | 58 | ||
| AK039840.1 | - | 1 - 2 | 146 | 58 | ||
| AK041567.1 | - | 1 - 2 | 126 | 58 | ||
| AK088274.1 | - | 1 - 2 | 145 | 58 | ||
| AK166608.1 | - | 1 - 2 | 128 | 58 | ||
| BC021748.1 | - | 1 - 2 | 134 | 3586619 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013517.1, AK166608.1, BC021748.1, NM_175190.5 transcripts.
Set Membership: |
|

Swi5
Genomic Map