Assay Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030112.2 | NP_084388.2 | 13 - 14 | 1698 | 69 | |
| GenBank mRNA | AK135633.1 | - | 7 - 8 | 766 | 69 | |
| BC117848.1 | - | 13 - 14 | 1550 | 40093717 | 69 | |
| BC117849.1 | - | 13 - 14 | 1550 | 40093724 | 69 | |
| CN532066.1 | - | 3 - 4 | 214 | 69 | ||
| CN839001.1 | - | 5 - 6 | 461 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rtf1
Genomic Map