Assay Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030112.2 | NP_084388.2 | 14 - 15 | 1753 | 68 | |
| GenBank mRNA | CN532066.1 | - | 4 - 5 | 269 | 68 | |
| CN839001.1 | - | 6 - 7 | 516 | 68 | ||
More Information
Set Membership: |
|

Rtf1
Genomic Map