Assay Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030112.2 | NP_084388.2 | 1 - 2 | 211 | 61 | |
| GenBank mRNA | BC038032.1 | - | 1 - 2 | 184 | 61 | |
| BC048535.1 | - | 1 - 2 | 195 | 61 | ||
| BC117848.1 | - | 1 - 2 | 63 | 40093717 | 61 | |
| BC117849.1 | - | 1 - 2 | 63 | 40093724 | 61 | |
| BY270385.1 | - | 1 - 2 | 205 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC117848.1, BC117849.1 transcripts.
Set Membership: |
|

Rtf1
Genomic Map