Assay Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030112.2 | NP_084388.2 | 10 - 11 | 1388 | 97 | |
| GenBank mRNA | AK135633.1 | - | 4 - 5 | 456 | 97 | |
| BC117848.1 | - | 10 - 11 | 1240 | 40093717 | 97 | |
| BC117849.1 | - | 10 - 11 | 1240 | 40093724 | 97 | |
| CN839001.1 | - | 2 - 3 | 151 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rtf1
Genomic Map