Assay Details
| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019696.2 | NP_062670.2 | 4 - 5 | 679 | 79 | |
| GenBank mRNA | AF077738.1 | - | 4 - 5 | 624 | 79 | |
| BC003713.1 | - | 4 - 5 | 612 | 3593211 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm1
Genomic Map