Assay Details
| Gene Symbol: Idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130884.4 | NP_570954.1 | 1 - 2 | 69 | 84 | |
| GenBank mRNA | AK046059.1 | - | 1 - 2 | 55 | 84 | |
| AY035212.1 | - | 1 - 2 | 50 | 84 | ||
| BC009022.1 | - | 1 - 2 | 58 | 3257667 | 84 | |
| BY336999.1 | - | 1 - 2 | 69 | 84 | ||
| DN176034.1 | - | 1 - 2 | 35 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idh3b
Genomic Map