Assay Details
| Gene Symbol: Ctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013256.2 | NP_001013274.2 | 7 - 8 | 1422 | 68 | |
| NM_001143790.1 | NP_001137262.1 | 7 - 8 | 1422 | 68 | ||
| NM_001281464.1 | NP_001268393.1 | 7 - 8 | 1293 | 68 | ||
| NM_001281465.1 | NP_001268394.1 | 7 - 8 | 1406 | 68 | ||
| GenBank mRNA | AK038651.1 | - | 7 - 8 | 1209 | 68 | |
| AK044055.1 | - | 7 - 8 | 1201 | 68 | ||
| AK145919.1 | - | 7 - 8 | 1237 | 68 | ||
| AK165906.1 | - | 7 - 8 | 1222 | 68 | ||
| BC060629.1 | - | 3 - 4 | 472 | 6847619 | 68 | |
| BC066078.1 | - | 7 - 8 | 1237 | 68 | ||
| BC072625.1 | - | 7 - 8 | 1250 | 68 | ||
| BC147495.1 | - | 7 - 8 | 1220 | 9056325 | 68 | |
| BC147497.1 | - | 7 - 8 | 1220 | 9056354 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038651.1 transcripts.
Set Membership: |
|

Ctc1
Genomic Map