Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025626.4 | NP_079902.1 | 1 - 2 | 178 | 126 | |
| GenBank mRNA | AK013932.2 | - | 1 - 2 | 143 | 127 | |
| AK019815.1 | - | 1 - 2 | 90 | 126 | ||
| AK032875.1 | - | 1 - 2 | 179 | 126 | ||
| AK168759.1 | - | 1 - 2 | 144 | 126 | ||
| BC021353.1 | - | 1 - 2 | 144 | 5006672 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013932.2, AK019815.1, AK032875.1, BC021353.1, NM_025626.4 transcripts.
Set Membership: |
|

Fam107b
Genomic Map