Assay Details
| Gene Symbol: Ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009832.2 | NP_033962.2 | 6 - 7 | 737 | 80 | |
| GenBank mRNA | AF060517.1 | - | 4 - 5 | 506 | 80 | |
| AK089386.1 | - | 6 - 7 | 674 | 80 | ||
| AK146711.1 | - | 6 - 7 | 674 | 80 | ||
| AK154679.1 | - | 6 - 7 | 674 | 80 | ||
| AK154765.1 | - | 6 - 7 | 674 | 80 | ||
| BC027297.1 | - | 6 - 7 | 632 | 3986609 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnk
Genomic Map