Assay Details
| Gene Symbol: Ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009832.2 | NP_033962.2 | 8 - 9 | 969 | 69 | |
| GenBank mRNA | AK089386.1 | - | 8 - 9 | 906 | 69 | |
| AK146711.1 | - | 8 - 9 | 906 | 69 | ||
| AK154679.1 | - | 8 - 9 | 906 | 69 | ||
| AK154765.1 | - | 8 - 9 | 906 | 69 | ||
| BC027297.1 | - | 8 - 9 | 864 | 3986609 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnk
Genomic Map