Assay Details
| Gene Symbol: Ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009832.2 | NP_033962.2 | 9 - 10 | 1245 | 100 | |
| GenBank mRNA | AK146711.1 | - | 9 - 10 | 1182 | 100 | |
| AK154679.1 | - | 9 - 10 | 1182 | 100 | ||
| BC027297.1 | - | 9 - 10 | 1140 | 3986609 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnk
Genomic Map